Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613552 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSAP - gamma-secretase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017439.3 | 2619 | UTR 3 | NP_059135.2 | |||
XM_011516325.2 | 2619 | UTR 3 | XP_011514627.1 | |||
XM_011516326.2 | 2619 | UTR 3 | XP_011514628.1 | |||
XM_011516327.2 | 2619 | UTR 3 | XP_011514629.1 | |||
XM_011516330.2 | 2619 | UTR 3 | XP_011514632.1 | |||
XM_011516331.2 | 2619 | Intron | XP_011514633.1 | |||
XM_011516332.2 | 2619 | Intron | XP_011514634.1 | |||
XM_017012343.1 | 2619 | UTR 3 | XP_016867832.1 | |||
XM_017012344.1 | 2619 | UTR 3 | XP_016867833.1 | |||
XM_017012345.1 | 2619 | UTR 3 | XP_016867834.1 | |||
XM_017012346.1 | 2619 | UTR 3 | XP_016867835.1 | |||
XM_017012347.1 | 2619 | UTR 3 | XP_016867836.1 | |||
XM_017012348.1 | 2619 | UTR 3 | XP_016867837.1 | |||
XM_017012349.1 | 2619 | UTR 3 | XP_016867838.1 | |||
XM_017012350.1 | 2619 | Intron | XP_016867839.1 | |||
XM_017012351.1 | 2619 | Intron | XP_016867840.1 |
Set Membership: |
HapMap |