Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601767 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HIP1 - huntingtin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243198.2 | 3440 | UTR 3 | NP_001230127.1 | |||
NM_005338.6 | 3440 | UTR 3 | NP_005329.3 | |||
XM_005250304.2 | 3440 | UTR 3 | XP_005250361.1 | |||
XM_005250305.2 | 3440 | UTR 3 | XP_005250362.1 | |||
XM_011516116.2 | 3440 | Intron | XP_011514418.1 | |||
XM_017012099.1 | 3440 | UTR 3 | XP_016867588.1 |
PMS2P3 - PMS1 homolog 2, mismatch repair system component pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |