Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615904 | ||||||||||||||||||||||||||||||||
Literature Links: |
PCMTD1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PCMTD1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286782.1 | 3391 | UTR 3 | NP_001273711.1 | |||
NM_001286783.1 | 3391 | UTR 3 | NP_001273712.1 | |||
NM_052937.3 | 3391 | UTR 3 | NP_443169.2 | |||
XM_005251146.2 | 3391 | Intron | XP_005251203.1 | |||
XM_011517444.2 | 3391 | UTR 3 | XP_011515746.1 | |||
XM_017013017.1 | 3391 | UTR 3 | XP_016868506.1 |
PXDNL - peroxidasin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |