Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171650 MIM: 600811 MIM: 602423 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACP2 - acid phosphatase 2, lysosomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302489.1 | 989 | Intron | NP_001289418.1 | |||
NM_001302490.1 | 989 | Intron | NP_001289419.1 | |||
NM_001302491.1 | 989 | Intron | NP_001289420.1 | |||
NM_001302492.1 | 989 | Intron | NP_001289421.1 | |||
NM_001610.3 | 989 | Intron | NP_001601.1 |
DDB2 - damage specific DNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000107.2 | 989 | UTR 3 | NP_000098.1 | |||
NM_001300734.1 | 989 | UTR 3 | NP_001287663.1 |
NR1H3 - nuclear receptor subfamily 1 group H member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |