Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608489 | ||||||||||||||||||||
Literature Links: |
GATS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GATS - GATS, stromal antigen 3 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178831.6 | 1306 | Intron | NP_849153.3 |
STAG3 - stromal antigen 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282716.1 | 1306 | Missense Mutation | ATG,GTG | M,V 396 | NP_001269645.1 | |
NM_001282717.1 | 1306 | Missense Mutation | ATG,GTG | M,V 396 | NP_001269646.1 | |
NM_001282718.1 | 1306 | Missense Mutation | ATG,GTG | M,V 338 | NP_001269647.1 | |
NM_012447.3 | 1306 | Missense Mutation | ATG,GTG | M,V 396 | NP_036579.2 |