Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602658 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01537 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01537 - long intergenic non-protein coding RNA 1537 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE2A - phosphodiesterase 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143839.3 | 4118 | UTR 3 | NP_001137311.1 | |||
NM_001146209.2 | 4118 | UTR 3 | NP_001139681.1 | |||
NM_001243784.1 | 4118 | UTR 3 | NP_001230713.1 | |||
NM_002599.4 | 4118 | UTR 3 | NP_002590.1 | |||
XM_005274040.2 | 4118 | UTR 3 | XP_005274097.1 | |||
XM_011545096.2 | 4118 | UTR 3 | XP_011543398.1 | |||
XM_017017910.1 | 4118 | UTR 3 | XP_016873399.1 |