Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616820 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ30679 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLJ30679 - uncharacterized protein FLJ30679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOXC2-AS1 - FOXC2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTHFSD - methenyltetrahydrofolate synthetase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159377.1 | Intron | NP_001152849.1 | ||||
NM_001159378.1 | Intron | NP_001152850.1 | ||||
NM_001159379.1 | Intron | NP_001152851.1 | ||||
NM_001159380.1 | Intron | NP_001152852.1 | ||||
NM_022764.2 | Intron | NP_073601.2 |
Set Membership: |
HapMap |