Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605799 MIM: 614062 | ||||||||||||||||||||||||||||||||
Literature Links: |
AMN PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
AMN - amnion associated transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030943.3 | 6562 | Intron | NP_112205.2 |
CDC42BPB - CDC42 binding protein kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006035.3 | 6562 | UTR 3 | NP_006026.3 | |||
XM_005268227.1 | 6562 | UTR 3 | XP_005268284.1 | |||
XM_005268228.1 | 6562 | UTR 3 | XP_005268285.1 | |||
XM_005268229.1 | 6562 | UTR 3 | XP_005268286.1 | |||
XM_005268230.4 | 6562 | Intron | XP_005268287.1 | |||
XM_011537387.1 | 6562 | Intron | XP_011535689.1 |
Set Membership: |
HapMap |