Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601153 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHIT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FHIT - fragile histidine triad | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166243.2 | 3519 | Intron | NP_001159715.1 | |||
NM_001320899.1 | 3519 | UTR 3 | NP_001307828.1 | |||
NM_001320900.1 | 3519 | UTR 3 | NP_001307829.1 | |||
NM_001320901.1 | 3519 | UTR 3 | NP_001307830.1 | |||
NM_002012.3 | 3519 | Intron | NP_002003.1 | |||
XM_017005880.1 | 3519 | UTR 3 | XP_016861369.1 | |||
XM_017005881.1 | 3519 | UTR 3 | XP_016861370.1 | |||
XM_017005882.1 | 3519 | UTR 3 | XP_016861371.1 | |||
XM_017005883.1 | 3519 | Intron | XP_016861372.1 | |||
XM_017005884.1 | 3519 | Intron | XP_016861373.1 | |||
XM_017005885.1 | 3519 | UTR 3 | XP_016861374.1 | |||
XM_017005886.1 | 3519 | UTR 3 | XP_016861375.1 |