Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606455 MIM: 613504 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R13B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PPP1R13B - protein phosphatase 1 regulatory subunit 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015316.2 | Intron | NP_056131.2 | ||||
XM_005267487.4 | Intron | XP_005267544.1 | ||||
XM_011536593.2 | Intron | XP_011534895.1 | ||||
XM_017021116.1 | Intron | XP_016876605.1 | ||||
XM_017021117.1 | Intron | XP_016876606.1 | ||||
XM_017021118.1 | Intron | XP_016876607.1 | ||||
XM_017021119.1 | Intron | XP_016876608.1 |
ZFYVE21 - zinc finger FYVE-type containing 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |