Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RD3L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RD3L - retinal degeneration 3-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDRD9 - tudor domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153046.2 | Intron | NP_694591.2 | ||||
XM_005267309.3 | Intron | XP_005267366.1 | ||||
XM_006720019.3 | Intron | XP_006720082.1 | ||||
XM_006720020.3 | Intron | XP_006720083.1 | ||||
XM_011536397.2 | Intron | XP_011534699.1 | ||||
XM_011536398.2 | Intron | XP_011534700.1 | ||||
XM_011536400.2 | Intron | XP_011534702.1 | ||||
XM_011536401.2 | Intron | XP_011534703.1 | ||||
XM_011536402.2 | Intron | XP_011534704.1 | ||||
XM_017020955.1 | Intron | XP_016876444.1 |
Set Membership: |
HapMap |