Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607640 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATXN7 - ataxin 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD6 - proteasome 26S subunit, non-ATPase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271779.1 | Intron | NP_001258708.1 | ||||
NM_001271780.1 | Intron | NP_001258709.1 | ||||
NM_001271781.1 | Intron | NP_001258710.1 | ||||
NM_014814.2 | Intron | NP_055629.1 | ||||
XM_005265619.1 | Intron | XP_005265676.1 | ||||
XM_011534288.1 | Intron | XP_011532590.1 | ||||
XM_017007569.1 | Intron | XP_016863058.1 |
PSMD6-AS2 - PSMD6 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |