Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTAAGTGGCAACTATATACTTGA[C/T]GCTGCCATTTCTTCTTTTTTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610670 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2U1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP2U1 - cytochrome P450 family 2 subfamily U member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183075.2 | Intron | NP_898898.1 | ||||
XM_005262717.2 | Intron | XP_005262774.1 | ||||
XM_005262720.2 | Intron | XP_005262777.1 |
LOC101929595 - uncharacterized LOC101929595 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |