Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153245 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LEF1 - lymphoid enhancer binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130713.2 | Intron | NP_001124185.1 | ||||
NM_001130714.2 | Intron | NP_001124186.1 | ||||
NM_001166119.1 | Intron | NP_001159591.1 | ||||
NM_016269.4 | Intron | NP_057353.1 | ||||
XM_005263046.3 | Intron | XP_005263103.1 | ||||
XM_005263047.1 | Intron | XP_005263104.1 | ||||
XM_005263048.1 | Intron | XP_005263105.1 | ||||
XM_006714233.1 | Intron | XP_006714296.1 |
Set Membership: |
Validated |