Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609042 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OPN5 - opsin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181744.3 | Intron | NP_859528.1 | ||||
XM_017010409.1 | Intron | XP_016865898.1 | ||||
XM_017010410.1 | Intron | XP_016865899.1 | ||||
XM_017010411.1 | Intron | XP_016865900.1 | ||||
XM_017010412.1 | Intron | XP_016865901.1 | ||||
XM_017010413.1 | Intron | XP_016865902.1 | ||||
XM_017010414.1 | Intron | XP_016865903.1 | ||||
XM_017010415.1 | Intron | XP_016865904.1 | ||||
XM_017010416.1 | Intron | XP_016865905.1 |
Set Membership: |
HapMap |