Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608761 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC5A7 - solute carrier family 5 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305005.2 | Intron | NP_001291934.1 | ||||
NM_001305006.2 | Intron | NP_001291935.1 | ||||
NM_001305007.2 | Intron | NP_001291936.1 | ||||
NM_021815.4 | Intron | NP_068587.1 | ||||
XM_011511580.2 | Intron | XP_011509882.1 | ||||
XM_017004628.1 | Intron | XP_016860117.1 | ||||
XM_017004629.1 | Intron | XP_016860118.1 |
Set Membership: |
HapMap |