Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 123691 MIM: 611245 | ||||||||||||||||||||
Literature Links: |
CRYZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRYZ - crystallin zeta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYW3 - tRNA-yW synthesizing protein 3 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162916.1 | 390 | Intron | NP_001156388.1 | |||
NM_138467.2 | 390 | Nonsense Mutation | GAA,TAA | E,* 101 | NP_612476.1 | |
XM_006710347.1 | 390 | Nonsense Mutation | GAA,TAA | E,* 101 | XP_006710410.1 |