Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605077 MIM: 609917 | ||||||||||||||||||||
Literature Links: |
DMAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DMAP1 - DNA methyltransferase 1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034023.1 | 1028 | Intron | NP_001029195.1 | |||
NM_001034024.1 | 1028 | Intron | NP_001029196.1 | |||
NM_019100.4 | 1028 | Intron | NP_061973.1 | |||
XM_005271037.4 | 1028 | Intron | XP_005271094.1 | |||
XM_005271039.1 | 1028 | Intron | XP_005271096.1 | |||
XM_005271040.1 | 1028 | Intron | XP_005271097.1 | |||
XM_006710771.3 | 1028 | Intron | XP_006710834.1 | |||
XM_011541784.2 | 1028 | Intron | XP_011540086.1 | |||
XM_011541785.2 | 1028 | Intron | XP_011540087.1 | |||
XM_017001807.1 | 1028 | Intron | XP_016857296.1 | |||
XM_017001808.1 | 1028 | Intron | XP_016857297.1 |
ERI3 - ERI1 exoribonuclease family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301698.1 | 1028 | UTR 3 | NP_001288627.1 | |||
NM_001301699.1 | 1028 | UTR 3 | NP_001288628.1 | |||
NM_001301700.1 | 1028 | UTR 3 | NP_001288629.1 | |||
NM_001301701.1 | 1028 | UTR 3 | NP_001288630.1 | |||
NM_024066.2 | 1028 | UTR 3 | NP_076971.1 | |||
XM_005271186.2 | 1028 | UTR 3 | XP_005271243.1 | |||
XM_006710891.3 | 1028 | UTR 3 | XP_006710954.1 | |||
XM_017002301.1 | 1028 | UTR 3 | XP_016857790.1 | |||
XM_017002302.1 | 1028 | Intron | XP_016857791.1 | |||
XM_017002303.1 | 1028 | UTR 3 | XP_016857792.1 | |||
XM_017002304.1 | 1028 | UTR 3 | XP_016857793.1 | |||
XM_017002305.1 | 1028 | UTR 3 | XP_016857794.1 |
ERI3-IT1 - ERI3 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |