Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609568 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP20 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP20 - Rho GTPase activating protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258415.1 | 4554 | UTR 3 | NP_001245344.1 | |||
NM_001258416.1 | 4554 | UTR 3 | NP_001245345.1 | |||
NM_001258417.1 | 4554 | UTR 3 | NP_001245346.1 | |||
NM_001258418.1 | 4554 | UTR 3 | NP_001245347.1 | |||
NM_020809.3 | 4554 | UTR 3 | NP_065860.2 | |||
XM_005271628.3 | 4554 | UTR 3 | XP_005271685.1 | |||
XM_006718893.3 | 4554 | UTR 3 | XP_006718956.1 |
Set Membership: |
HapMap |