Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609424 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RC3H1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RC3H1 - ring finger and CCCH-type domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300850.1 | 6392 | UTR 3 | NP_001287779.1 | |||
NM_001300851.1 | 6392 | UTR 3 | NP_001287780.1 | |||
NM_001300852.1 | 6392 | UTR 3 | NP_001287781.1 | |||
NM_172071.3 | 6392 | UTR 3 | NP_742068.1 | |||
XM_005244921.3 | 6392 | UTR 3 | XP_005244978.1 | |||
XM_011509231.2 | 6392 | UTR 3 | XP_011507533.1 |
Set Membership: |
HapMap Validated |