Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOL10 - nucleolar protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261392.1 | 1951 | UTR 3 | NP_001248321.1 | |||
NM_001261394.1 | 1951 | UTR 3 | NP_001248323.1 | |||
NM_024894.3 | 1951 | UTR 3 | NP_079170.2 | |||
XM_011510398.2 | 1951 | UTR 3 | XP_011508700.1 | |||
XM_011510399.2 | 1951 | UTR 3 | XP_011508701.1 | |||
XM_011510400.2 | 1951 | Intron | XP_011508702.1 | |||
XM_011510402.2 | 1951 | Intron | XP_011508704.1 | |||
XM_017004990.1 | 1951 | UTR 3 | XP_016860479.1 | |||
XM_017004991.1 | 1951 | UTR 3 | XP_016860480.1 |