Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609154 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASCL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASCL3 - achaete-scute family bHLH transcription factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM9B - TMEM9 domain family member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286094.1 | 1615 | UTR 3 | NP_001273023.1 | |||
NM_001286095.1 | 1615 | UTR 3 | NP_001273024.1 | |||
NM_020644.2 | 1615 | UTR 3 | NP_065695.1 |