Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609191 MIM: 140750 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKIP1 - A-kinase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206646.1 | 172 | Missense Mutation | AAG,AGG | K,R 23 | NP_001193575.1 | |
NM_001206647.1 | 172 | Missense Mutation | AAG,AGG | K,R 23 | NP_001193576.1 | |
NM_001206648.1 | 172 | Missense Mutation | AAG,AGG | K,R 23 | NP_001193577.1 | |
NM_020642.3 | 172 | Missense Mutation | AAG,AGG | K,R 23 | NP_065693.2 | |
XM_017018011.1 | 172 | Missense Mutation | AAG,AGG | K,R 23 | XP_016873500.1 | |
XM_017018012.1 | 172 | Missense Mutation | AAG,AGG | K,R 23 | XP_016873501.1 |
C11orf16 - chromosome 11 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |