Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613922 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LIPK - lipase family member K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080518.1 | Intron | NP_001073987.1 | ||||
XM_011540073.2 | Intron | XP_011538375.1 | ||||
XM_011540075.1 | Intron | XP_011538377.1 | ||||
XM_011540077.1 | Intron | XP_011538379.1 | ||||
XM_011540082.1 | Intron | XP_011538384.1 | ||||
XM_017016545.1 | Intron | XP_016872034.1 | ||||
XM_017016546.1 | Intron | XP_016872035.1 | ||||
XM_017016547.1 | Intron | XP_016872036.1 | ||||
XM_017016548.1 | Intron | XP_016872037.1 |
Set Membership: |
HapMap |