Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUCO PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SUCO - SUN domain containing ossification factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282750.1 | Intron | NP_001269679.1 | ||||
NM_001282751.1 | Intron | NP_001269680.1 | ||||
NM_014283.4 | Intron | NP_055098.1 | ||||
NM_016227.3 | Intron | NP_057311.3 | ||||
XM_006711374.2 | Intron | XP_006711437.1 | ||||
XM_006711375.2 | Intron | XP_006711438.1 | ||||
XM_006711376.1 | Intron | XP_006711439.1 |
Set Membership: |
HapMap |