Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607081 MIM: 185880 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAPBPL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TAPBPL - TAP binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018009.4 | 908 | Intron | NP_060479.3 | |||
XM_005253700.3 | 908 | Intron | XP_005253757.1 | |||
XM_017019545.1 | 908 | Intron | XP_016875034.1 | |||
XM_017019546.1 | 908 | Intron | XP_016875035.1 |
VAMP1 - vesicle associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297438.1 | 908 | Intron | NP_001284367.1 | |||
NM_014231.4 | 908 | Intron | NP_055046.1 | |||
NM_016830.3 | 908 | Intron | NP_058439.1 | |||
NM_199245.2 | 908 | UTR 3 | NP_954740.1 |
Set Membership: |
HapMap |