Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600654 MIM: 602161 MIM: 612487 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EMC9 - ER membrane protein complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7703 - microRNA 7703 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME1 - proteasome activator subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME2 - proteasome activator subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002818.2 | 798 | Intron | NP_002809.2 | |||
XM_006720213.1 | 798 | UTR 5 | XP_006720276.1 |
RNF31 - ring finger protein 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310332.1 | 798 | Intron | NP_001297261.1 | |||
NM_017999.4 | 798 | Intron | NP_060469.4 |
Set Membership: |
HapMap |