Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602443 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPNPEP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
XPNPEP1 - X-prolyl aminopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167604.1 | 2141 | UTR 3 | NP_001161076.1 | |||
NM_001324128.1 | 2141 | UTR 3 | NP_001311057.1 | |||
NM_001324131.1 | 2141 | UTR 3 | NP_001311060.1 | |||
NM_001324132.1 | 2141 | UTR 3 | NP_001311061.1 | |||
NM_001324133.1 | 2141 | UTR 3 | NP_001311062.1 | |||
NM_001324134.1 | 2141 | UTR 3 | NP_001311063.1 | |||
NM_001324135.1 | 2141 | UTR 3 | NP_001311064.1 | |||
NM_001324136.1 | 2141 | UTR 3 | NP_001311065.1 | |||
NM_020383.3 | 2141 | UTR 3 | NP_065116.3 | |||
XM_011540134.1 | 2141 | UTR 3 | XP_011538436.1 | |||
XM_017016613.1 | 2141 | UTR 3 | XP_016872102.1 | |||
XM_017016614.1 | 2141 | UTR 3 | XP_016872103.1 | |||
XM_017016615.1 | 2141 | UTR 3 | XP_016872104.1 |
Set Membership: |
HapMap |