Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZSCAN32 - zinc finger and SCAN domain containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284527.1 | 2615 | UTR 3 | NP_001271456.1 | |||
NM_001284528.1 | 2615 | UTR 3 | NP_001271457.1 | |||
NM_001284529.1 | 2615 | UTR 3 | NP_001271458.1 | |||
NM_001324340.1 | 2615 | UTR 3 | NP_001311269.1 | |||
NM_001324341.1 | 2615 | UTR 3 | NP_001311270.1 | |||
NM_001324342.1 | 2615 | UTR 3 | NP_001311271.1 | |||
NM_001324343.1 | 2615 | UTR 3 | NP_001311272.1 | |||
NM_001324344.1 | 2615 | UTR 3 | NP_001311273.1 | |||
NM_001324345.1 | 2615 | UTR 3 | NP_001311274.1 | |||
NM_001324346.1 | 2615 | UTR 3 | NP_001311275.1 | |||
NM_017810.3 | 2615 | UTR 3 | NP_060280.2 | |||
XM_005255403.2 | 2615 | UTR 3 | XP_005255460.1 | |||
XM_011522555.2 | 2615 | UTR 3 | XP_011520857.1 | |||
XM_017023371.1 | 2615 | UTR 3 | XP_016878860.1 | |||
XM_017023372.1 | 2615 | UTR 3 | XP_016878861.1 | |||
XM_017023373.1 | 2615 | UTR 3 | XP_016878862.1 |
Set Membership: |
HapMap Validated |