Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300663 MIM: 303631 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATG4A - autophagy related 4A cysteine peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COL4A6 - collagen type IV alpha 6 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287758.1 | 5853 | UTR 3 | NP_001274687.1 | |||
NM_001287759.1 | 5853 | UTR 3 | NP_001274688.1 | |||
NM_001287760.1 | 5853 | UTR 3 | NP_001274689.1 | |||
NM_001847.3 | 5853 | UTR 3 | NP_001838.2 | |||
NM_033641.3 | 5853 | UTR 3 | NP_378667.1 | |||
XM_006724617.3 | 5853 | UTR 3 | XP_006724680.1 | |||
XM_011530852.2 | 5853 | Intron | XP_011529154.1 | |||
XM_011530853.2 | 5853 | Intron | XP_011529155.1 | |||
XM_011530854.2 | 5853 | Intron | XP_011529156.1 |
Set Membership: |
HapMap |