Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602871 MIM: 609771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPL - periplakin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002705.4 | 5394 | UTR 3 | NP_002696.3 | |||
XM_006720902.3 | 5394 | UTR 3 | XP_006720965.1 | |||
XM_017023374.1 | 5394 | UTR 3 | XP_016878863.1 | |||
XM_017023375.1 | 5394 | UTR 3 | XP_016878864.1 |
UBN1 - ubinuclein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079514.2 | 5394 | Intron | NP_001072982.1 | |||
NM_001288656.1 | 5394 | Intron | NP_001275585.1 | |||
XM_005255277.3 | 5394 | Intron | XP_005255334.1 | |||
XM_005255278.3 | 5394 | Intron | XP_005255335.1 | |||
XM_005255279.3 | 5394 | Intron | XP_005255336.1 | |||
XM_011522465.1 | 5394 | Intron | XP_011520767.1 | |||
XM_011522466.2 | 5394 | Intron | XP_011520768.1 | |||
XM_011522467.1 | 5394 | Intron | XP_011520769.1 | |||
XM_017023189.1 | 5394 | Intron | XP_016878678.1 | |||
XM_017023190.1 | 5394 | Intron | XP_016878679.1 | |||
XM_017023191.1 | 5394 | Intron | XP_016878680.1 | |||
XM_017023192.1 | 5394 | Intron | XP_016878681.1 |