Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300012 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMARCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SMARCA1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282874.1 | Intron | NP_001269803.1 | ||||
NM_001282875.1 | Intron | NP_001269804.1 | ||||
NM_003069.4 | Intron | NP_003060.2 | ||||
XM_005262461.2 | Intron | XP_005262518.1 | ||||
XM_005262462.2 | Intron | XP_005262519.1 | ||||
XM_006724782.2 | Intron | XP_006724845.1 | ||||
XM_017029750.1 | Intron | XP_016885239.1 | ||||
XM_017029751.1 | Intron | XP_016885240.1 |
Set Membership: |
HapMap Validated |