Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300242 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC25A14 - solute carrier family 25 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282195.1 | Intron | NP_001269124.1 | ||||
NM_001282196.1 | Intron | NP_001269125.1 | ||||
NM_001282197.1 | Intron | NP_001269126.1 | ||||
NM_001282198.1 | Intron | NP_001269127.1 | ||||
XM_005262489.4 | Intron | XP_005262546.1 | ||||
XM_011531402.2 | Intron | XP_011529704.1 | ||||
XM_011531403.1 | Intron | XP_011529705.1 | ||||
XM_017029936.1 | Intron | XP_016885425.1 | ||||
XM_017029937.1 | Intron | XP_016885426.1 | ||||
XM_017029938.1 | Intron | XP_016885427.1 | ||||
XM_017029939.1 | Intron | XP_016885428.1 | ||||
XM_017029940.1 | Intron | XP_016885429.1 | ||||
XM_017029941.1 | Intron | XP_016885430.1 | ||||
XM_017029942.1 | Intron | XP_016885431.1 | ||||
XM_017029943.1 | Intron | XP_016885432.1 |
Set Membership: |
HapMap |