Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606568 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZTFL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LZTFL1 - leucine zipper transcription factor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276378.1 | 3919 | UTR 3 | NP_001263307.1 | |||
NM_001276379.1 | 3919 | UTR 3 | NP_001263308.1 | |||
NM_020347.3 | 3919 | UTR 3 | NP_065080.1 | |||
XM_006713207.2 | 3919 | UTR 3 | XP_006713270.2 | |||
XM_011533838.2 | 3919 | Intron | XP_011532140.1 | |||
XM_017006644.1 | 3919 | Intron | XP_016862133.1 | |||
XM_017006645.1 | 3919 | Intron | XP_016862134.1 |
Set Membership: |
HapMap |