Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604017 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT6 - beta-1,4-galactosyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004775.3 | 1796 | UTR 3 | NP_004766.2 | |||
XM_005258387.4 | 1796 | UTR 3 | XP_005258444.1 | |||
XM_006722578.3 | 1796 | UTR 3 | XP_006722641.1 | |||
XM_006722579.3 | 1796 | UTR 3 | XP_006722642.1 | |||
XM_017026088.1 | 1796 | UTR 3 | XP_016881577.1 | |||
XM_017026089.1 | 1796 | UTR 3 | XP_016881578.1 | |||
XM_017026090.1 | 1796 | UTR 3 | XP_016881579.1 | |||
XM_017026091.1 | 1796 | UTR 3 | XP_016881580.1 | |||
XM_017026092.1 | 1796 | UTR 3 | XP_016881581.1 |