Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607056 MIM: 612846 | ||||||||||||||||||||
Literature Links: |
IMPG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IMPG2 - interphotoreceptor matrix proteoglycan 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SENP7 - SUMO1/sentrin specific peptidase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077203.2 | 2932 | Missense Mutation | AAT,GAT | N,D 941 | NP_001070671.1 | |
NM_001282801.1 | 2932 | Missense Mutation | AAT,GAT | N,D 940 | NP_001269730.1 | |
NM_001282802.1 | 2932 | Missense Mutation | AAT,GAT | N,D 973 | NP_001269731.1 | |
NM_001282803.1 | 2932 | Missense Mutation | AAT,GAT | N,D 842 | NP_001269732.1 | |
NM_001282804.1 | 2932 | Intron | NP_001269733.1 | |||
NM_020654.4 | 2932 | Missense Mutation | AAT,GAT | N,D 1006 | NP_065705.3 |