Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189901 MIM: 611832 | ||||||||||||||||||||
Literature Links: |
GCFC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCFC2 - GC-rich sequence DNA-binding factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201334.1 | 4081 | UTR 3 | NP_001188263.1 | |||
NM_001201335.1 | 4081 | Intron | NP_001188264.1 | |||
NM_003203.4 | 4081 | UTR 3 | NP_003194.3 | |||
XM_005264520.3 | 4081 | UTR 3 | XP_005264577.1 | |||
XM_011533074.2 | 4081 | UTR 3 | XP_011531376.1 | |||
XM_011533075.1 | 4081 | Intron | XP_011531377.1 | |||
XM_017004787.1 | 4081 | Intron | XP_016860276.1 | |||
XM_017004788.1 | 4081 | Intron | XP_016860277.1 |
MRPL19 - mitochondrial ribosomal protein L19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014763.3 | 4081 | Intron | NP_055578.2 |