Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602190 | ||||||||||||||||||||
Literature Links: |
EPHA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPHA7 - EPH receptor A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288629.1 | 4920 | UTR 3 | NP_001275558.1 | |||
NM_001288630.1 | 4920 | Intron | NP_001275559.1 | |||
NM_004440.3 | 4920 | UTR 3 | NP_004431.1 | |||
XM_005248669.2 | 4920 | UTR 3 | XP_005248726.1 | |||
XM_005248671.2 | 4920 | Intron | XP_005248728.1 | |||
XM_017010365.1 | 4920 | UTR 3 | XP_016865854.1 | |||
XM_017010366.1 | 4920 | Intron | XP_016865855.1 |