Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603893 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TANK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TANK - TRAF family member associated NFKB activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199135.1 | Intron | NP_001186064.1 | ||||
NM_004180.2 | Intron | NP_004171.2 | ||||
NM_133484.1 | Intron | NP_597841.1 | ||||
XM_005246207.3 | Intron | XP_005246264.2 | ||||
XM_017003093.1 | Intron | XP_016858582.1 | ||||
XM_017003094.1 | Intron | XP_016858583.1 | ||||
XM_017003095.1 | Intron | XP_016858584.1 | ||||
XM_017003096.1 | Intron | XP_016858585.1 | ||||
XM_017003097.1 | Intron | XP_016858586.1 |