Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAACATAAAAAATAAAATAAGCA[A/G]ACTGTCTGAGGGGTTTATATAAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAP - fibroblast activation protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291807.1 | 2479 | UTR 3 | NP_001278736.1 | |||
NM_004460.3 | 2479 | UTR 3 | NP_004451.2 | |||
XM_011510796.2 | 2479 | UTR 3 | XP_011509098.1 | |||
XM_011510797.2 | 2479 | Intron | XP_011509099.1 | |||
XM_017003585.1 | 2479 | Intron | XP_016859074.1 |
LOC101929532 - uncharacterized LOC101929532 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |