Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603512 | ||||||||||||||||||||
Literature Links: |
RNGTT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNGTT - RNA guanylyltransferase and 5'-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286426.1 | 2790 | UTR 3 | NP_001273355.1 | |||
NM_001286428.1 | 2790 | UTR 3 | NP_001273357.1 | |||
NM_003800.4 | 2790 | UTR 3 | NP_003791.3 | |||
XM_017011401.1 | 2790 | UTR 3 | XP_016866890.1 | |||
XM_017011402.1 | 2790 | Intron | XP_016866891.1 |