Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM209A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM209A - family with sequence similarity 209 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012971.3 | 386 | Missense Mutation | AAA,AAT | K,N 102 | NP_001012989.2 |
FAM209B - family with sequence similarity 209 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCNT7 - glucosaminyl (N-acetyl) transferase family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080615.1 | 386 | UTR 5 | NP_542182.1 |
RTFDC1 - replication termination factor 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |