Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600378 MIM: 614918 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IMMT - inner membrane mitochondrial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100169.1 | 2170 | Silent Mutation | GCC,GCT | A,A 594 | NP_001093639.1 | |
NM_001100170.1 | 2170 | Silent Mutation | GCC,GCT | A,A 584 | NP_001093640.1 | |
NM_006839.2 | 2170 | Silent Mutation | GCC,GCT | A,A 595 | NP_006830.2 | |
XM_005264110.3 | 2170 | Intron | XP_005264167.1 | |||
XM_005264113.1 | 2170 | Intron | XP_005264170.1 | |||
XM_005264114.1 | 2170 | Intron | XP_005264171.1 | |||
XM_011532497.2 | 2170 | Intron | XP_011530799.1 | |||
XM_017003194.1 | 2170 | Intron | XP_016858683.1 | |||
XM_017003195.1 | 2170 | Intron | XP_016858684.1 |
PTCD3 - pentatricopeptide repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD94 - small nucleolar RNA, C/D box 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |