Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606718 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGXB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMGXB3 - HMG-box containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC26A2 - solute carrier family 26 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIGD6 - tigger transposable element derived 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243253.1 | 3547 | UTR 3 | NP_001230182.1 | |||
NM_030953.3 | 3547 | UTR 3 | NP_112215.1 |
Set Membership: |
HapMap |