Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180071 MIM: 608886 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDE6A - phosphodiesterase 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000440.2 | 3137 | UTR 3 | NP_000431.2 | |||
XM_011537650.2 | 3137 | UTR 3 | XP_011535952.1 | |||
XM_011537651.2 | 3137 | UTR 3 | XP_011535953.1 | |||
XM_011537652.1 | 3137 | UTR 3 | XP_011535954.1 | |||
XM_011537653.2 | 3137 | UTR 3 | XP_011535955.1 | |||
XM_011537654.2 | 3137 | UTR 3 | XP_011535956.1 | |||
XM_017009572.1 | 3137 | UTR 3 | XP_016865061.1 |
PPARGC1B - PPARG coactivator 1 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |