Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606157 MIM: 612489 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PANK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PANK2 - pantothenate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324191.1 | 3138 | Intron | NP_001311120.1 | |||
NM_001324192.1 | 3138 | Intron | NP_001311121.1 | |||
NM_001324193.1 | 3138 | Intron | NP_001311122.1 | |||
NM_024960.5 | 3138 | Intron | NP_079236.3 | |||
NM_153638.3 | 3138 | Intron | NP_705902.2 | |||
NM_153640.3 | 3138 | Intron | NP_705904.1 |
RNF24 - ring finger protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134337.2 | 3138 | UTR 3 | NP_001127809.1 | |||
NM_001134338.2 | 3138 | UTR 3 | NP_001127810.1 | |||
NM_001321749.1 | 3138 | UTR 3 | NP_001308678.1 | |||
NM_007219.4 | 3138 | UTR 3 | NP_009150.1 | |||
XM_011529145.2 | 3138 | UTR 3 | XP_011527447.1 | |||
XM_011529146.2 | 3138 | UTR 3 | XP_011527448.1 | |||
XM_011529147.2 | 3138 | Intron | XP_011527449.1 | |||
XM_017027620.1 | 3138 | UTR 3 | XP_016883109.1 | |||
XM_017027621.1 | 3138 | UTR 3 | XP_016883110.1 | |||
XM_017027622.1 | 3138 | Intron | XP_016883111.1 |
Set Membership: |
HapMap Validated |