Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600810 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PLCB4 - phospholipase C beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000933.3 | Intron | NP_000924.3 | ||||
NM_001172646.1 | Intron | NP_001166117.1 | ||||
NM_182797.2 | Intron | NP_877949.2 | ||||
XM_005260724.2 | Intron | XP_005260781.1 | ||||
XM_005260728.2 | Intron | XP_005260785.1 | ||||
XM_006723568.3 | Intron | XP_006723631.1 | ||||
XM_006723569.2 | Intron | XP_006723632.1 | ||||
XM_011529254.2 | Intron | XP_011527556.1 | ||||
XM_017027880.1 | Intron | XP_016883369.1 | ||||
XM_017027881.1 | Intron | XP_016883370.1 | ||||
XM_017027882.1 | Intron | XP_016883371.1 | ||||
XM_017027883.1 | Intron | XP_016883372.1 | ||||
XM_017027884.1 | Intron | XP_016883373.1 | ||||
XM_017027885.1 | Intron | XP_016883374.1 | ||||
XM_017027886.1 | Intron | XP_016883375.1 |
Set Membership: |
HapMap Validated |