Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTTGTAATAATTCTGAGCACA[C/T]GGAGCAAACTGTAAACTGGTCATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615870 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFT27 - intraflagellar transport 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177701.2 | Intron | NP_001171172.1 | ||||
NM_006860.4 | Intron | NP_006851.1 | ||||
XM_006724106.2 | Intron | XP_006724169.1 | ||||
XM_017028540.1 | Intron | XP_016884029.1 |
LOC105373021 - uncharacterized LOC105373021 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |