Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 602496 MIM: 180370 | ||||||||||||||||||||||||||
Literature Links: |
MPST PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MPST - mercaptopyruvate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX33 - testis expressed 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TST - thiosulfate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270483.1 | 726 | Missense Mutation | GAG,GGG | E,G 228 | NP_001257412.1 | |
NM_003312.5 | 726 | Missense Mutation | GAG,GGG | E,G 228 | NP_003303.2 |
Set Membership: |
HapMap |