Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606610 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSFL1C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NSFL1C - NSFL1 cofactor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRPB2 - signal regulatory protein beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122962.1 | 2259 | UTR 3 | NP_001116434.1 | |||
NM_001134836.1 | 2259 | UTR 3 | NP_001128308.1 | |||
XM_005260708.4 | 2259 | Intron | XP_005260765.1 | |||
XM_005260709.3 | 2259 | Intron | XP_005260766.1 | |||
XM_011529221.2 | 2259 | Intron | XP_011527523.1 | |||
XM_011529223.2 | 2259 | Intron | XP_011527525.1 | |||
XM_011529224.2 | 2259 | Intron | XP_011527526.1 | |||
XM_011529225.2 | 2259 | Intron | XP_011527527.1 |
Set Membership: |
HapMap |